Assay Details
| Gene Symbol: Lrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080926.1 | NP_001074395.1 | 5 - 6 | 758 | 79 | |
| NR_033496.1 | 6 - 7 | 1136 | 79 | |||
| GenBank mRNA | AJ312058.1 | - | 5 - 6 | 604 | 79 | |
| D85463.1 | - | 6 - 7 | 1134 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp8
Genomic Map