Assay Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053093.1 | NP_444323.1 | 3 - 4 | 448 | 67 | |
| GenBank mRNA | AF235035.1 | - | 3 - 4 | 448 | 67 | |
| AF515827.1 | - | 3 - 4 | 282 | 67 | ||
| BC119426.1 | - | 3 - 4 | 428 | 40039772 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037691 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037685 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037462 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037683 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037688 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124064 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161972 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28037700 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02400581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161972 | - | - |
| Whole Mouse Genome G4122A | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474498 | NM_053093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474498 | AF235035.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116495 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161972 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319859 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09606150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09595821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09319854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037685 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037683 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037691 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037462 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28037460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28037670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09579463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09467196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09319852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09671583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09579462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09356820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09319855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09356822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09446025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09319859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09660268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09726509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09022340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09595817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09124127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09579468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09225496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28037730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380511 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420458_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380506 | - | - |
| Mouse Genome 430 2.0 Array | 1420458_at | - | - |
| Mouse Genome 430 2.0 Array | 1420458_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93670_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93670_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100013525.mm.1 | NM_053093.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013525.mm.1 | NM_053093.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013525.mm.1 | TC1100001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013526.mm.1 | NM_053093.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013526.mm.1 | NM_053093.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013526.mm.1 | TC1100001457.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420458_PM_at | - | - |
| Mouse Expression Set 430 | 1420458_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420458_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420458_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420458_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420458_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 855933 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4730070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4730070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653965 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653965 | - | - |
| mouse-6 v1.1 (Illumina) | scl41014.4.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41014.4.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237288 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237288 | - | - |

Tac4
Genomic Map