Assay Details
Gene Symbol: Pcdhb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053138.2 | NP_444368.1 | 1 | 601 | 91 | |
GenBank mRNA | AK082435.1 | - | 1 | 603 | 91 | |
AY013774.1 | - | 1 | 547 | 91 | ||
BC138123.1 | - | 1 | 618 | 8861142 | 91 | |
BC138124.1 | - | 1 | 618 | 8861143 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425858 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143969 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P204809 | AK082435.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P204809 | AK082435.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264708 | BC138123.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264708 | BC138124.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264708 | AK082435.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264708 | AY013774.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264708 | NM_053138.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628294 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628296 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628305 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628321 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628312 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628307 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628314 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628302 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628303 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628309 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628300 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143969 | - | - |
Whole Mouse Genome G4122A | A_51_P264708 | BC138123.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P264708 | BC138124.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P264708 | AK082435.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P264708 | AY013774.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P264708 | NM_053138.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P204809 | AK082435.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P204809 | NM_053138.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108787 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143969 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13725506 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13725512 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13725526 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13725497 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13725520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13646487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628287 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13725514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628283 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10455097 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455096 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455095 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450316_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10455094 | - | - |
Mouse Genome 430 2.0 Array | 1450316_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450316_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93884_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93884_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800002845.mm.1 | NM_053138.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002845.mm.1 | NM_053138.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002845.mm.1 | TC1800000369.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450316_at | - | - |
Mouse Expression Set 430 | 1450316_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450316_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450316_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450316_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450316_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 512582 | - | - |
ABI Mouse Genome Survey Microarray | 656999 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102690435 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6840082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840082 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598879 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2598879 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2443913 | - | - |
mouse-6 v1.1 (Illumina) | scl0093884.2_291-S | - | - |
mouse-6 v1.1 (Illumina) | scl52061.1_698-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840082 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102690435 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093884.2_291-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840082 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102690435 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1239577 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2443913 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239577 | - | - |
