Assay Details
| Gene Symbol: Pcdhb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053145.2 | NP_444375.2 | 1 | 466 | 96 | |
| GenBank mRNA | AF326313.1 | - | 1 | 442 | 96 | |
| AK045399.1 | - | 1 | 467 | 96 | ||
| AK045527.1 | - | 1 | 440 | 96 | ||
| AK046137.1 | - | 1 | 459 | 96 | ||
| AY013782.1 | - | 1 | 307 | 96 | ||
| BC031895.1 | - | 1 | 401 | 5364116 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578390 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107329 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628667 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107329 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08968342 | - | - |
| Whole Mouse Genome G4122A | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | BC031895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK045527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK046137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AK045399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146760 | AF326313.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124073 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13973653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13973657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13959291 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13674811 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13973637 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13973655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13973666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13973629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13959291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14072313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13674813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628663 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455132 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449583_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455128 | - | - |
| Mouse Genome 430 2.0 Array | 1449583_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449583_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93891_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93891_at | - | - |
| Mouse Expression Set 430 | 1449583_at | - | - |
| Mouse Expression Set 430 | 1449583_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449583_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449583_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449583_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449583_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 651940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 870373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000870373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712974 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712974 | - | - |
| mouse-6 v1.1 (Illumina) | scl52055.1.28_104-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 870373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52055.1.28_104-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2979449 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2979449 | - | - |

Pcdhb20
Genomic Map