Assay Details
| Gene Symbol: Ces1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053200.2 | NP_444430.2 | 7 - 8 | 979 | 91 | |
| GenBank mRNA | AB023631.1 | - | 7 - 8 | 939 | 91 | |
| AF378751.1 | - | 7 - 8 | 954 | 91 | ||
| AK078879.1 | - | 7 - 8 | 957 | 91 | ||
| BC019198.1 | - | 7 - 8 | 951 | 5040523 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces1d
Genomic Map