Assay Details
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053248.2 | NP_444478.2 | 10 - 11 | 1338 | 88 | |
GenBank mRNA | AF235001.1 | - | 10 - 11 | 1305 | 88 | |
AF380353.1 | - | 10 - 11 | 1260 | 88 | ||
AK046718.1 | - | 10 - 11 | 1339 | 88 | ||
BC137650.1 | - | 10 - 11 | 1273 | 8860669 | 88 | |
BC137651.1 | - | 10 - 11 | 1273 | 8860670 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
