Assay Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054055.2 | NP_473396.1 | 7 - 8 | 1049 | 74 | |
| GenBank mRNA | AF306491.1 | - | 7 - 8 | 1048 | 74 | |
| AK085326.1 | - | 7 - 8 | 1047 | 74 | ||
| AK087518.1 | - | 7 - 8 | 1047 | 74 | ||
| AK128982.1 | - | 7 - 8 | 1046 | 74 | ||
| AK138344.1 | - | 7 - 8 | 1047 | 74 | ||
| AK157881.1 | - | 7 - 8 | 1018 | 74 | ||
| AK165355.1 | - | 7 - 8 | 1048 | 74 | ||
| BC026803.1 | - | 7 - 8 | 1035 | 4238769 | 74 | |
| BC028882.1 | - | 6 - 7 | 908 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a3
Genomic Map