Assay Details
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054095.2 | NP_473436.1 | 6 - 7 | 939 | 65 | |
| GenBank mRNA | AF411253.1 | - | 6 - 7 | 933 | 65 | |
| AK035168.1 | - | 4 - 5 | 314 | 65 | ||
| AK082727.1 | - | 6 - 7 | 549 | 65 | ||
| BC107384.1 | - | 7 - 8 | 534 | 65 | ||
| BC107385.1 | - | 7 - 8 | 498 | 65 | ||
| BC115857.1 | - | 7 - 8 | 471 | 40057629 | 65 | |
| BC115963.1 | - | 7 - 8 | 459 | 40057628 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007163 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706436 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AB041590.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AF151521.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AF172286.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AF176423.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AF232241.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AJ243895.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AK011149.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | AK140212.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | BC086635.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814590 | NM_010423.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161776 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177714 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109674 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26214089 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177714 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109674 | - | - |
| Whole Mouse Genome G4122A | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | AF411253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | AK035168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | AK082727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181341 | NM_054095.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116838 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109674 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06922197 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240207 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06922208 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06922188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P33007164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26214043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06685703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06685701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06922207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06685702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06749365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33007161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06724099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06749362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06872414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33007163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07010889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06749359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07240205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P33007162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26214105 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575885 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418881_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575880 | - | - |
| Mouse Genome 430 2.0 Array | 1418881_at | - | - |
| Mouse Genome 430 2.0 Array | 1418881_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117148_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117148_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011556.mm.1 | NM_054095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011556.mm.1 | NM_054095.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011556.mm.1 | TC0800001421.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418881_PM_at | - | - |
| Mouse Expression Set 430 | 1418881_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418881_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418881_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418881_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418881_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 449520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 610368 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000610368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694360 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694360 | - | - |
| mouse-6 v1.1 (Illumina) | scl33260.8.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 610368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33260.8.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610368 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694360 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694360 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33260.8.1_8-S | - | - |

Necab2
Genomic Map