Assay Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080463.3 | NP_536711.3 | 4 - 5 | 615 | 65 | |
GenBank mRNA | AF375885.1 | - | 4 - 5 | 584 | 65 | |
AK044629.1 | - | 4 - 5 | 593 | 65 | ||
AK081059.1 | - | 4 - 5 | 572 | 65 | ||
AK132301.1 | - | 4 - 5 | 570 | 65 | ||
AK135635.1 | - | 4 - 5 | 597 | 65 | ||
AK170038.1 | - | 4 - 5 | 570 | 65 | ||
BC046295.1 | - | 4 - 5 | 615 | 6485896 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
