| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400015841.mm.1 |
NM_007455.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015841.mm.1 |
TC1400002080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015843.mm.1 |
NM_007455.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015843.mm.1 |
TC1400002080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015844.mm.1 |
NM_007455.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015844.mm.1 |
TC1400002080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015850.mm.1 |
NM_007455.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015850.mm.1 |
TC1400002080.mm.1 |
MTA TC |