Assay Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007504.2 | NP_031530.2 | 20 - 21 | 3014 | 61 | |
| GenBank mRNA | AK132537.1 | - | 5 - 6 | 731 | 61 | |
| AK160497.1 | - | 3 - 4 | 303 | 61 | ||
| AY081946.1 | - | 20 - 21 | 3045 | 61 | ||
| BC036292.1 | - | 20 - 21 | 3014 | 4191741 | 61 | |
| X67140.1 | - | 4 - 5 | 427 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2a1
Genomic Map