Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600011515.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011515.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011515.mm.1 |
NM_007531.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011523.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011523.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011523.mm.1 |
NM_007531.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011525.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011525.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011525.mm.1 |
NM_007531.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011509.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011509.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011509.mm.1 |
NM_007531.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011508.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011508.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011508.mm.1 |
NM_007531.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011514.mm.1 |
TC0600001427.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600011514.mm.1 |
NM_007531.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600011514.mm.1 |
NM_007531.2 |
Refseq |