Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 20 - 21 | 2714 | 74 | |
| GenBank mRNA | AB119195.1 | - | 18 - 19 | 2212 | 74 | |
| AB221614.1 | - | 18 - 19 | 2219 | 74 | ||
| AK089660.1 | - | 8 - 9 | 1088 | 74 | ||
| AK146389.1 | - | 6 - 7 | 933 | 74 | ||
| AK159423.1 | - | 8 - 9 | 1145 | 74 | ||
| AY521462.1 | - | 18 - 19 | 2212 | 74 | ||
| AY521463.1 | - | 18 - 19 | 2212 | 74 | ||
| BC138535.1 | - | 19 - 20 | 2291 | 8861555 | 74 | |
| BC138536.1 | - | 19 - 20 | 2291 | 8861556 | 74 | |
| BC145317.1 | - | 20 - 21 | 2308 | 74 | ||
| M57999.1 | - | 20 - 21 | 2503 | 74 | ||
| S66656.1 | - | 8 - 9 | 1258 | 74 | ||
| S89033.1 | - | 8 - 9 | 1306 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map