Assay Details
Gene Symbol: Nptx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008730.2 | NP_032756.1 | 3 - 4 | 1129 | 74 | |
GenBank mRNA | AK140128.1 | - | 3 - 4 | 646 | 74 | |
BC093479.1 | - | 3 - 4 | 1054 | 30947054 | 74 | |
BC139018.1 | - | 3 - 4 | 1036 | 8862040 | 74 | |
BC139020.1 | - | 3 - 4 | 1036 | 8862042 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
