Assay Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285890.1 | NP_001272819.1 | 6 - 7 | 1065 | 67 | |
| NM_008800.2 | NP_032826.1 | 7 - 8 | 1082 | 67 | ||
| GenBank mRNA | AK004772.1 | - | 6 - 7 | 672 | 67 | |
| BC058531.1 | - | 7 - 8 | 1082 | 6826674 | 67 | |
| L01695.1 | - | 7 - 8 | 825 | 67 | ||
| M94538.1 | - | 1 - 2 | 70 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde1b
Genomic Map