Assay Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285890.1 | NP_001272819.1 | 13 - 14 | 1839 | 90 | |
| NM_008800.2 | NP_032826.1 | 14 - 15 | 1856 | 90 | ||
| GenBank mRNA | AK004772.1 | - | 13 - 14 | 1446 | 90 | |
| L01695.1 | - | 14 - 15 | 1599 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde1b
Genomic Map