Assay Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037551.1 | - | 9 - 10 | 2365 | 71 | |
| AK084527.1 | - | 8 - 9 | 1955 | 71 | ||
| AK169431.1 | - | 7 - 8 | 1654 | 71 | ||
| BC008519.1 | - | 8 - 9 | 1959 | 3592726 | 71 | |
| BC085502.1 | - | 8 - 9 | 1761 | 5720681 | 71 | |
| L06039.1 | - | 8 - 9 | 1844 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pecam1
Genomic Map