Assay Details
| Gene Symbol: Pml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008884.5 | NP_032910.3 | 5 - 6 | 1646 | 74 | |
| NM_178087.4 | NP_835188.2 | 6 - 7 | 1784 | 74 | ||
| GenBank mRNA | AK028044.1 | - | 6 - 7 | 1786 | 74 | |
| AK032799.1 | - | 4 - 5 | 664 | 74 | ||
| AK137538.1 | - | 6 - 7 | 1732 | 74 | ||
| AK156279.1 | - | 6 - 7 | 1944 | 74 | ||
| BC020990.1 | - | 6 - 7 | 1591 | 4188386 | 74 | |
| KJ650238.1 | - | 6 - 7 | 1688 | 74 | ||
| U33626.2 | - | 5 - 6 | 1543 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pml
Genomic Map