Assay Details
| Gene Symbol: Pml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008884.5 | NP_032910.3 | 6 - 7 | 1696 | 66 | |
| NM_178087.4 | NP_835188.2 | 7 - 8 | 1834 | 66 | ||
| GenBank mRNA | AK028044.1 | - | 7 - 8 | 1836 | 66 | |
| AK032799.1 | - | 5 - 6 | 714 | 66 | ||
| AK052305.1 | - | 7 - 8 | 1833 | 66 | ||
| AK137538.1 | - | 7 - 8 | 1782 | 66 | ||
| AK156279.1 | - | 7 - 8 | 1994 | 66 | ||
| BC020990.1 | - | 7 - 8 | 1641 | 4188386 | 66 | |
| KJ650238.1 | - | 7 - 8 | 1738 | 66 | ||
| U33626.2 | - | 6 - 7 | 1593 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pml
Genomic Map