Assay Details
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008902.3 | NP_032928.1 | 2 - 3 | 373 | 144 | |
GenBank mRNA | M95545.1 | - | 2 - 3 | 373 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
