Assay Details
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168693.1 | NP_001162164.1 | 5 - 6 | 690 | 96 | |
NM_008902.3 | NP_032928.1 | 4 - 5 | 689 | 96 | ||
GenBank mRNA | AK132616.1 | - | 5 - 6 | 631 | 96 | |
AK169457.1 | - | 3 - 4 | 780 | 96 | ||
M95545.1 | - | 4 - 5 | 689 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
