Assay Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008950.1 | NP_032976.1 | 3 - 4 | 173 | 76 | |
| GenBank mRNA | AK010505.1 | - | 3 - 4 | 193 | 76 | |
| AK135451.1 | - | 3 - 4 | 183 | 76 | ||
| AK145703.1 | - | 3 - 4 | 177 | 76 | ||
| BC030840.1 | - | 3 - 4 | 205 | 3994523 | 76 | |
| Z54219.1 | - | 3 - 4 | 173 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc5
Genomic Map