Assay Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008950.1 | NP_032976.1 | 5 - 6 | 559 | 91 | |
| GenBank mRNA | AK010505.1 | - | 5 - 6 | 579 | 91 | |
| AK012294.1 | - | 5 - 6 | 592 | 91 | ||
| AK135451.1 | - | 5 - 6 | 569 | 91 | ||
| AK145703.1 | - | 5 - 6 | 563 | 91 | ||
| BC004052.1 | - | 1 - 2 | 254 | 91 | ||
| BC030840.1 | - | 5 - 6 | 591 | 3994523 | 91 | |
| Z54219.1 | - | 5 - 6 | 559 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc5
Genomic Map