Assay Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008950.1 | NP_032976.1 | 8 - 9 | 975 | 112 | |
| GenBank mRNA | AK010505.1 | - | 8 - 9 | 995 | 112 | |
| AK012294.1 | - | 8 - 9 | 1008 | 112 | ||
| AK135451.1 | - | 8 - 9 | 985 | 112 | ||
| AK145703.1 | - | 8 - 9 | 979 | 112 | ||
| BC004052.1 | - | 4 - 5 | 670 | 112 | ||
| Z54219.1 | - | 8 - 9 | 975 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc5
Genomic Map