Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030184.mm.1 |
NM_008952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030184.mm.1 |
NM_008952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030184.mm.1 |
TC1100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030185.mm.1 |
NM_008952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030185.mm.1 |
NM_008952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030185.mm.1 |
TC1100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030183.mm.1 |
NM_008952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030183.mm.1 |
NM_008952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030183.mm.1 |
TC1100003276.mm.1 |
MTA TC |