Assay Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008969.4 | NP_032995.1 | 5 - 6 | 647 | 81 | |
| GenBank mRNA | AK046457.1 | - | 5 - 6 | 574 | 81 | |
| AK134163.1 | - | 5 - 6 | 533 | 81 | ||
| AK138432.1 | - | 5 - 6 | 586 | 81 | ||
| AK142741.1 | - | 5 - 6 | 574 | 81 | ||
| AK159907.1 | - | 5 - 6 | 648 | 81 | ||
| AK160886.1 | - | 5 - 6 | 566 | 81 | ||
| AK163609.1 | - | 5 - 6 | 574 | 81 | ||
| AK167357.1 | - | 4 - 5 | 525 | 81 | ||
| AK170414.1 | - | 5 - 6 | 542 | 81 | ||
| AK170664.1 | - | 5 - 6 | 574 | 81 | ||
| BC005573.1 | - | 5 - 6 | 564 | 3968101 | 81 | |
| BC023322.1 | - | 5 - 6 | 556 | 3968136 | 81 | |
| CT010314.1 | - | 4 - 5 | 510 | 81 | ||
| M34141.1 | - | 5 - 6 | 546 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptgs1
Genomic Map