| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500014670.mm.1 |
TC0500001620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014670.mm.1 |
NM_009599.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014671.mm.1 |
TC0500001620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014671.mm.1 |
NM_009599.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014668.mm.1 |
TC0500001620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014672.mm.1 |
TC0500001620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014672.mm.1 |
NM_009599.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014662.mm.1 |
TC0500001620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014662.mm.1 |
NM_009599.4 |
Refseq |