Assay Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009673.2 | NP_033803.1 | 9 - 10 | 965 | 146 | |
| GenBank mRNA | AK147740.1 | - | 9 - 10 | 744 | 146 | |
| AK152185.1 | - | 9 - 10 | 800 | 146 | ||
| AK153388.1 | - | 9 - 10 | 746 | 146 | ||
| AK153476.1 | - | 9 - 10 | 729 | 146 | ||
| D63423.1 | - | 9 - 10 | 703 | 146 | ||
| U29396.1 | - | 9 - 10 | 722 | 146 | ||
More Information
Set Membership: |
|

Anxa5
Genomic Map