Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700021022.mm.1 |
NM_001110832.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021022.mm.1 |
NM_001110832.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021022.mm.1 |
NM_010913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021022.mm.1 |
NM_010913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021022.mm.1 |
TC1700002225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021026.mm.1 |
NM_001110832.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021026.mm.1 |
NM_001110832.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021026.mm.1 |
NM_010913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021026.mm.1 |
NM_010913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021026.mm.1 |
TC1700002225.mm.1 |
MTA TC |