Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 14 - 15 | 1705 | 66 | |
| NM_011031.2 | NP_035161.2 | 14 - 15 | 1711 | 66 | ||
| GenBank mRNA | AK087663.1 | - | 1 - 2 | 1429 | 66 | |
| AK142689.1 | - | 6 - 7 | 400 | 66 | ||
| AK145535.1 | - | 14 - 15 | 1632 | 66 | ||
| BC018411.1 | - | 14 - 15 | 1617 | 3157481 | 66 | |
| CD563665.1 | - | 7 - 8 | 569 | 66 | ||
| U16163.1 | - | 13 - 14 | 1598 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha2
Genomic Map