Assay Details
| Gene Symbol: Pabpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011033.2 | NP_035163.1 | 1 | 1669 | 97 | |
| GenBank mRNA | AK077154.1 | - | 1 | 1669 | 97 | |
| BC051134.1 | - | 1 | 1649 | 6704229 | 97 | |
| X75959.1 | - | 1 | 1645 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pabpc2
Genomic Map