Assay Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011116.1 | NP_035246.1 | 6 - 7 | 668 | 119 | |
| GenBank mRNA | AF026124.1 | - | 6 - 7 | 668 | 119 | |
| AK153508.1 | - | 4 - 5 | 357 | 119 | ||
| BC068170.1 | - | 6 - 7 | 665 | 119 | ||
| BC076586.1 | - | 6 - 7 | 732 | 30604186 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pld3
Genomic Map