Assay Details
Gene Symbol: Prl4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011165.3 | NP_035295.1 | 1 - 2 | 84 | 59 | |
GenBank mRNA | AF011383.1 | - | 1 - 2 | 85 | 59 | |
AF015562.1 | - | 1 - 2 | 84 | 59 | ||
AK141285.1 | - | 1 - 2 | 96 | 59 | ||
AK164009.1 | - | 1 - 2 | 96 | 59 | ||
BC052218.1 | - | 1 - 2 | 85 | 30017285 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86649 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144099 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134376 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | BC052218.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AF011383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AF015562.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AK141285.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | BC052218.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AF011383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AF015562.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904300 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904297 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28904281 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134376 | - | - |
Whole Mouse Genome G4122A | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | BC052218.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | NM_011165.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AF011383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AF015562.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455166 | AK141285.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138878 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102874 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134376 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10615717 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10615720 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10615707 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10615705 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10615741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28904246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10615711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904248 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11065276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10615709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10958715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11065281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10615739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10800357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10445654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28904246 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404346 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404345 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404344 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404342 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404343 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448572_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404341 | - | - |
Mouse Genome 430 2.0 Array | 1448572_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448572_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19110_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19110_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001911.mm.1 | NM_011165.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001911.mm.1 | NM_011165.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001911.mm.1 | TC1300000308.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001912.mm.1 | NM_011165.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001912.mm.1 | NM_011165.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001912.mm.1 | TC1300000308.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448572_PM_at | - | - |
Mouse Expression Set 430 | 1448572_at | - | - |
Murine Genome U74v2 Set | 103378_PM_at | - | - |
Murine Genome U74v2 Set | 103378_at | - | - |
Mouse Genome 430A 2.0 Array | 1448572_at | - | - |
Mouse Genome 430A 2.0 Array | 1448572_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448572_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448572_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 323918 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6980427 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980427 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607776 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2607776 | - | - |
mouse-6 v1.1 (Illumina) | scl44961.5.1_198-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980427 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44961.5.1_198-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980427 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2607776 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2607776 | - | - |
