| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146120.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146120.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146121.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146122.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146122.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146123.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_001146123.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_011179.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
NM_011179.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003688.mm.1 |
TC1000000547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146120.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146120.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146121.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146122.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146122.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146123.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146123.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146124.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_001146124.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_011179.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
NM_011179.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003690.mm.1 |
TC1000000547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146120.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146120.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146121.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146121.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146122.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146122.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146123.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_001146123.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_011179.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
NM_011179.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003687.mm.1 |
TC1000000547.mm.1 |
MTA TC |