Assay Details
| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011309.3 | NP_035439.1 | 1 - 2 | 132 | 82 | |
| GenBank mRNA | AF087687.1 | - | 1 - 2 | 99 | 82 | |
| AK002721.1 | - | 1 - 2 | 73 | 82 | ||
| AK012578.1 | - | 1 - 2 | 132 | 82 | ||
| AK017279.1 | - | 1 - 2 | 68 | 82 | ||
| AK168822.1 | - | 1 - 2 | 68 | 82 | ||
| BC005590.1 | - | 1 - 2 | 204 | 3964547 | 82 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF087687.1,AK002721.1,AK012578.1,AK017279.1,AK168822.1,BC005590.1,NM_011309.3 transcripts.
Set Membership: |
|

S100a1
Genomic Map