Assay Details
| Gene Symbol: S100a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011312.2 | NP_035442.1 | 1 - 2 | 160 | 104 | |
| GenBank mRNA | AF087469.1 | - | 1 - 2 | 160 | 104 | |
| AI327447.1 | - | 1 - 2 | 160 | 104 | ||
| BC117108.1 | - | 1 - 2 | 159 | 40126427 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060333 | NM_011312.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060333 | AI327447.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703633 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140700 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060333 | NM_011312.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060333 | AI327447.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140700 | - | - |
| Whole Mouse Genome G4122A | A_51_P108812 | NM_011312.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108812 | AF087469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108812 | AI327447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P108812 | BC117108.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111237 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140700 | - | - |
| Agilent Mouse cDNA Microarray (G4104A) | 315361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03062376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03062371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02828767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02565101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02828768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03052295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03062372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02565104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02935072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03052285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493818 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449927_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493816 | - | - |
| Mouse Genome 430 2.0 Array | 1449927_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449927_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20199_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20199_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006305.mm.1 | NM_011312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006305.mm.1 | NM_011312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006305.mm.1 | TC0300000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006304.mm.1 | NM_011312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006304.mm.1 | NM_011312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006304.mm.1 | TC0300000808.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006306.mm.1 | NM_011312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006306.mm.1 | NM_011312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006306.mm.1 | TC0300000808.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449927_at | - | - |
| Mouse Expression Set 430 | 1449927_PM_at | - | - |
| Murine Genome U74v2 Set | 95301_at | - | - |
| Murine Genome U74v2 Set | 95301_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449927_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449927_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449927_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449927_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 539507 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2749169 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2749169 | - | - |
| mouse-6 v1.1 (Illumina) | scl22943.2.1_50-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22943.2.1_50-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2749169 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749169 | - | - |

S100a5
Genomic Map