Assay Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011786.2 | NP_035916.2 | 6 - 7 | 813 | 65 | |
| GenBank mRNA | AK141003.1 | - | 5 - 6 | 3579 | 65 | |
| BC116258.1 | - | 6 - 7 | 812 | 40093733 | 65 | |
| BC116259.1 | - | 6 - 7 | 812 | 40093735 | 65 | |
| Y14695.1 | - | 6 - 7 | 813 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aloxe3
Genomic Map