Assay Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011786.2 | NP_035916.2 | 12 - 13 | 1711 | 128 | |
| GenBank mRNA | BC116258.1 | - | 12 - 13 | 1710 | 40093733 | 128 |
| BC116259.1 | - | 12 - 13 | 1710 | 40093735 | 128 | |
| Y14695.1 | - | 12 - 13 | 1711 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aloxe3
Genomic Map