Assay Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286743.1 | NP_001273672.1 | 4 - 5 | 778 | 79 | |
| NM_001286744.1 | NP_001273673.1 | 4 - 5 | 684 | 79 | ||
| NM_011861.3 | NP_035991.1 | 4 - 5 | 743 | 79 | ||
| NM_178365.4 | NP_848142.1 | 4 - 5 | 761 | 79 | ||
| GenBank mRNA | AK043967.1 | - | 4 - 5 | 747 | 79 | |
| AK158686.1 | - | 4 - 5 | 781 | 79 | ||
| BC014698.1 | - | 4 - 5 | 632 | 4527708 | 79 | |
| X85124.1 | - | 4 - 5 | 652 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin1
Genomic Map