Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300005909.mm.1 |
NM_013628.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005909.mm.1 |
NM_013628.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005909.mm.1 |
TC1300000908.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005908.mm.1 |
NM_013628.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005908.mm.1 |
NM_013628.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005908.mm.1 |
TC1300000908.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005910.mm.1 |
NM_013628.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005910.mm.1 |
NM_013628.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005910.mm.1 |
TC1300000908.mm.1 |
MTA TC |