| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163467.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163468.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163469.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_001163469.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_013629.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
NM_013629.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008684.mm.1 |
TC0300001070.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163467.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163468.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163469.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_001163469.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_013629.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
NM_013629.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008683.mm.1 |
TC0300001070.mm.1 |
MTA TC |