Assay Details
| Gene Symbol: Ate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029895.3 | NP_001025066.1 | 5 - 6 | 979 | 86 | |
| NM_001136054.2 | NP_001129526.1 | 5 - 6 | 711 | 86 | ||
| NM_001271343.1 | NP_001258272.1 | 5 - 6 | 979 | 86 | ||
| NM_013799.3 | NP_038827.2 | 5 - 6 | 711 | 86 | ||
| GenBank mRNA | AK077471.1 | - | 1 - 2 | 130 | 86 | |
| AK138277.1 | - | 5 - 6 | 630 | 86 | ||
| AK148442.1 | - | 5 - 6 | 679 | 86 | ||
| AK167591.1 | - | 5 - 6 | 671 | 86 | ||
| BC042601.1 | - | 5 - 6 | 670 | 4503326 | 86 | |
| BC050948.1 | - | 5 - 6 | 661 | 6826691 | 86 | |
| DQ093639.1 | - | 5 - 6 | 571 | 86 | ||
| DQ093640.1 | - | 5 - 6 | 571 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ate1
Genomic Map