Assay Details
| Gene Symbol: Pstpip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013831.4 | NP_038859.3 | 813 | 73 | ||
| GenBank mRNA | AK132418.1 | - | 795 | 73 | ||
| AK154284.1 | - | 780 | 73 | |||
| AK154724.1 | - | 9 - 10 | 819 | 73 | ||
| AK157994.1 | - | 9 - 10 | 797 | 73 | ||
| BC002123.1 | - | 794 | 3494357 | 73 | ||
| BC057654.1 | - | 755 | 4460817 | 73 | ||
| CT010308.1 | - | 9 - 10 | 645 | 73 | ||
| Y18101.1 | - | 9 - 10 | 645 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pstpip2
Genomic Map