Assay Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016784.3 | NP_058064.2 | 5 - 6 | 511 | 60 | |
| GenBank mRNA | AF044334.1 | - | 4 - 5 | 403 | 60 | |
| AK075993.1 | - | 5 - 6 | 499 | 60 | ||
| AK076007.1 | - | 5 - 6 | 489 | 60 | ||
| AK077197.1 | - | 5 - 6 | 512 | 60 | ||
| AK080068.1 | - | 5 - 6 | 518 | 60 | ||
| BC006750.1 | - | 5 - 6 | 481 | 3709134 | 60 | |
| BC052895.1 | - | 5 - 6 | 469 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plrg1
Genomic Map