| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300011623.mm.1 |
NM_001111324.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011623.mm.1 |
NM_017464.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011623.mm.1 |
NM_017464.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011623.mm.1 |
TC1300001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011626.mm.1 |
NM_001111324.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011626.mm.1 |
TC1300001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011627.mm.1 |
NM_001111324.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011627.mm.1 |
TC1300001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011625.mm.1 |
NM_001111324.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011625.mm.1 |
TC1300001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300011628.mm.1 |
NM_001111324.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300011628.mm.1 |
TC1300001861.mm.1 |
MTA TC |