Assay Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018873.2 | NP_061361.2 | 8 - 9 | 1940 | 63 | |
| GenBank mRNA | AF040944.1 | - | 8 - 9 | 1942 | 63 | |
| AK129422.1 | - | 3 - 4 | 1097 | 63 | ||
| U59873.1 | - | 8 - 9 | 1941 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srcin1
Genomic Map