Assay Details
| Gene Symbol: Paf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019458.3 | NP_062331.2 | 10 - 11 | 1068 | 72 | |
| GenBank mRNA | AB041615.1 | - | 10 - 11 | 1086 | 72 | |
| AK017762.1 | - | 10 - 11 | 1138 | 72 | ||
| AK134857.1 | - | 10 - 11 | 1081 | 72 | ||
| AK148538.1 | - | 10 - 11 | 1103 | 72 | ||
| AK152778.1 | - | 10 - 11 | 1084 | 72 | ||
| BC010317.1 | - | 9 - 10 | 1112 | 72 | ||
| BC029843.1 | - | 10 - 11 | 1068 | 2651081 | 72 | |
| BC083337.1 | - | 10 - 11 | 1049 | 5701517 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paf1
Genomic Map