Assay Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019798.5 | NP_062772.3 | 4 - 5 | 865 | 102 | |
| NM_183408.3 | NP_899668.1 | 9 - 10 | 1373 | 102 | ||
| GenBank mRNA | AF208021.2 | - | 4 - 5 | 537 | 102 | |
| AJ297396.1 | - | 4 - 5 | 537 | 102 | ||
| AK153657.1 | - | 9 - 10 | 1378 | 102 | ||
| AK160843.1 | - | 8 - 9 | 807 | 102 | ||
| BC027224.1 | - | 9 - 10 | 102 | |||
| BC044786.2 | - | 8 - 9 | 102 | |||
| BC139405.1 | - | 9 - 10 | 1213 | 8862427 | 102 | |
| BC139408.1 | - | 9 - 10 | 1213 | 8862430 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4a
Genomic Map