Assay Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019800.4 | NP_062774.2 | 4 - 5 | 933 | 66 | |
| GenBank mRNA | AB030038.1 | - | 4 - 5 | 926 | 66 | |
| AF216223.1 | - | 4 - 5 | 410 | 66 | ||
| AK002783.1 | - | 4 - 5 | 847 | 66 | ||
| AK077729.1 | - | 4 - 5 | 921 | 66 | ||
| AK160271.1 | - | 4 - 5 | 852 | 66 | ||
| AK165356.1 | - | 4 - 5 | 934 | 66 | ||
| AK171757.1 | - | 4 - 5 | 848 | 66 | ||
| BC047276.1 | - | 4 - 5 | 673 | 6487551 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acp6
Genomic Map