Assay Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019800.4 | NP_062774.2 | 6 - 7 | 1145 | 77 | |
| GenBank mRNA | AB030038.1 | - | 6 - 7 | 1138 | 77 | |
| AF216223.1 | - | 6 - 7 | 622 | 77 | ||
| AK002783.1 | - | 6 - 7 | 1059 | 77 | ||
| AK077729.1 | - | 6 - 7 | 1133 | 77 | ||
| AK160271.1 | - | 6 - 7 | 1064 | 77 | ||
| AK165356.1 | - | 6 - 7 | 1146 | 77 | ||
| AK171757.1 | - | 6 - 7 | 1060 | 77 | ||
| BC047276.1 | - | 6 - 7 | 885 | 6487551 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acp6
Genomic Map